Skip to Content
MilliporeSigma
1,3,5-Triaza-7-phosphaadamantane 97%

1,3,5-Triaza-7-phosphaadamantane

1,3,5-Triaza-7-phosphaadamantane 97%
Synonym(s):
1,3,5-Triaza-7-phosphatricyclo[3.3.1.13.7]decane, NSC 266642, PTA
Empirical Formula (Hill Notation):
C6H12N3P
CAS Number:
Molecular Weight:
157.15

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing