Skip to Content
MilliporeSigma
1-(2-Pyridyl)piperazine 98%

1-(2-Pyridyl)piperazine

1-(2-Pyridyl)piperazine 98%
Synonym(s):
2-Piperazinopyridine
Empirical Formula (Hill Notation):
C9H13N3
CAS Number:
Molecular Weight:
163.22
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing