Skip to Content
MilliporeSigma
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine ≥97%

1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine

1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine ≥97%
Synonym(s):
1,2-Dihexadecanoyl-sn-glycero-3-phosphoethanolamine, 3-sn-Phosphatidylethanolamine, 1,2-dipalmitoyl, L-α-Phosphatidylethanolamine, dipalmitoyl, L-β,γ-Dipalmitoyl-α-cephalin, DPPE, Dipalmitoyl phosphatidylethanolamine, Hexadecanoic acid (1R)-1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, PE(16:0/16:0)
Empirical Formula (Hill Notation):
C37H74NO8P
CAS Number:
Molecular Weight:
691.96

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing