Skip to Content
MilliporeSigma
1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine synthetic, ≥99%

1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine

1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine synthetic, ≥99%
Synonym(s):
1,2-Ditetradecanoyl-sn-glycero-3-phosphoethanolamine, 3-sn-Phosphatidylethanolamine, 1,2-dimyristoyl, L-β,γ-Dimyristoyl-α-cephalin, DMPE, Dimyristoyl-L-α-phosphatidylethanolamine, PE(14:0/14:0), Tetradecanoic acid (1R)-1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester
Empirical Formula (Hill Notation):
C33H66NO8P
CAS Number:
Molecular Weight:
635.85

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing