Skip to Content
MilliporeSigma
1,2-Dichloropropane Pharmaceutical Secondary Standard; Certified Reference Material

1,2-Dichloropropane

1,2-Dichloropropane Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Propylene dichloride, 1,2-Dichloropropane
Linear Formula:
CH3CHClCH2Cl
CAS Number:
Molecular Weight:
112.99

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique