Skip to Content
MilliporeSigma
1,2-Bis(chlorodimethylsilyl)ethane 96%

1,2-Bis(chlorodimethylsilyl)ethane

1,2-Bis(chlorodimethylsilyl)ethane 96%
Synonym(s):
1,1,4,4-Tetramethyl-1,4-dichloro-disilethylene, 2,5-Dichloro-2,5-dimethyl-2,5-disilahexane
Linear Formula:
[-CH2Si(CH3)2Cl]2
CAS Number:
Molecular Weight:
215.27
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing