Skip to Content
MilliporeSigma
1,2,3,4-Butanetetracarboxylic acid 99%

1,2,3,4-Butanetetracarboxylic acid

1,2,3,4-Butanetetracarboxylic acid 99%
Synonym(s):
1,2,3,4-Butanetetracarboxylic acid
Linear Formula:
[-CH(CO2H)CH2CO2H]2
CAS Number:
Molecular Weight:
234.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing