1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate
Synonym(s):
DiI
Empirical Formula (Hill Notation):
C59H97ClN2O4
CAS Number:
Molecular Weight:
933.87
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Chemical Composition
Application
Showing 1-2 of 2