Skip to Content
MilliporeSigma
[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), complex with dichloromethane

[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), complex with dichloromethane

[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), complex with dichloromethane
Synonym(s):
Pd(dppf)Cl2 · CH2Cl2, Pd(dppf)Cl2 · DCM, [1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II) (1:1)
Empirical Formula (Hill Notation):
C35H30Cl4FeP2Pd
CAS Number:
Molecular Weight:
816.64

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-1 of 1