Skip to Content
MilliporeSigma
11ß,16a,17,21-Tetrahydroxypregna-1,4-dien-3,20-dione Pharmaceutical Analytical Impurity (PAI)

11ß,16a,17,21-Tetrahydroxypregna-1,4-dien-3,20-dione

11ß,16a,17,21-Tetrahydroxypregna-1,4-dien-3,20-dione Pharmaceutical Analytical Impurity (PAI)
Synonym(s):
11β,16α,17,21-Tetrahydroxypregna-1,4-dien-3,20-dione, 16α-Hydroxyprednisolone
Empirical Formula (Hill Notation):
C21H28O6
CAS Number:
Molecular Weight:
376.44

Products

Formula Weight
Manufacturer
Agency
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1