Product Name
tert-Butyl (tetrahydro-2-oxo-3-furanyl)carbamate 98%
Product Number
467049
Product Brand
aldrich
CAS Number
132957-40-5
Molecular Weight
201.22
Test
Specification
Appearance (Form)Powder or Crystals
Infrared spectrumConforms to Structure
Carbon52.4 - 55.1 %
Nitrogen6.7 - 7.2 %
Purity (GC)≥97.5 %