Product Name
Methacrylic acid 99%
Product Number
395374
Product Brand
aldrich
CAS Number
79-41-4
Molecular Weight
86.09
Test
Specification
APPEARANCECOLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE.
TITRATION98.5% - 101.5% (WITH NAOH)
COLOR TEST20 APHA (MAXIMUM)
 REVISED SEPTEMBER 15, 2005 BTR