Product Name
(R)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride 98%
Product Number
369667
Product Brand
aldrich
CAS Number
39637-99-5
Molecular Weight
252.62
Test
Specification
APPEARANCECOLORLESS LIQUID OR WHITE SOLID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
 OR
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
OPTICAL ROTATION-137(+/- 2) DEGREES (C=6.4%, CCL4)
 OR
OPTICAL ROTATION-132(+/- 2) DEGREES (C=6%, CHCL3)
ENANTIOMERIC EXCESS97.5% (MINIMUM)
GAS LIQUID97.5% (MINIMUM)
 REVISED JANUARY 30, 2005 RJM