Product Name
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid 97%
Product Number
238813
Product Brand
aldrich
CAS Number
53188-07-1
Molecular Weight
250.29
Test
Specification
APPEARANCE (COLOR)White to Light Beige
APPEARANCE (FORM)Powder or Chunks
TITRATION (T) NAOH 0.1M96.5 - 103.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE