Skip to Content
MilliporeSigma
All Photos(1)

Documents

V900662

Sigma-Aldrich

3,5-Pyridinedicarboxylic acid

Vetec, reagent grade, 98%

Synonym(s):

Dinicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO4
CAS Number:
Molecular Weight:
167.12
Beilstein:
131640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mp

>300 °C (lit.)

SMILES string

OC(=O)c1cncc(c1)C(O)=O

InChI

1S/C7H5NO4/c9-6(10)4-1-5(7(11)12)3-8-2-4/h1-3H,(H,9,10)(H,11,12)

InChI key

MPFLRYZEEAQMLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao-Yan Zhang et al.
Journal of chromatography. A, 1383, 18-24 (2015-01-27)
Three Ag(I)-organic frameworks, [Ag5(pydc)2(CN)]n, {[Ag4(pydc)2]CH3CN}n, and [Ag(4,4'-bpy)NO3]n, were synthesized and embedded into silica gels to form metal-organic-framework (MOF)-embedded gels for the microextraction of polybrominated diphenyl ethers (PBDEs) in soils. Despite the great differences in the structures of the organic ligands
Yihui Cui et al.
The Journal of physiology, 593(13), 2833-2849 (2015-04-16)
Although learning can arise from few or even a single trial, synaptic plasticity is commonly assessed under prolonged activation. Here, we explored the existence of rapid responsiveness of synaptic plasticity at corticostriatal synapses in a major synaptic learning rule, spike-timing-dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service