Skip to Content
MilliporeSigma
All Photos(3)

Documents

V900338

Sigma-Aldrich

L-Proline

≥99%,Vetec

Synonym(s):

(S)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
80810
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

product name

L-Proline, Vetec, reagent grade, ≥99%

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

color

white

mp

228 °C (dec.) (lit.)

SMILES string

OC(=O)[C@@H]1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1

InChI key

ONIBWKKTOPOVIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel R Knight et al.
Applied and environmental microbiology, 81(1), 119-123 (2014-10-19)
Clostridium difficile is an important enteric pathogen of humans and the cause of diarrhea and enteritis in neonatal pigs. Outside Australia, prevalence in piglets can be up to 73%, with a single PCR ribotype (RT), 078, predominating. We investigated the
Marie Couturier et al.
The Journal of biological chemistry, 288(20), 14624-14635 (2013-04-06)
The microbial deconstruction of the plant cell wall is a key biological process that is of increasing importance with the development of a sustainable biofuel industry. The glycoside hydrolase families GH5 (PaMan5A) and GH26 (PaMan26A) endo-β-1,4-mannanases from the coprophilic ascomycete
Salvatore Petta et al.
Hepatology (Baltimore, Md.), 59(5), 1692-1705 (2014-04-03)
We assessed the cost-effectiveness of sofosbuvir (SOF)-based triple therapy (TT) compared with boceprevir (BOC)- and telaprevir (TVR)-based TT in untreated genotype 1 (G1) chronic hepatitis C (CHC) patients discriminated according to IL28B genotype, severity of liver fibrosis, and G1 subtype.
Michael P Manns et al.
Gastroenterology, 147(2), 366-376 (2014-04-15)
MK-5172 is an inhibitor of the hepatitis C virus (HCV) nonstructural protein 3/4A protease; MK-5172 is taken once daily and has a higher potency and barrier to resistance than licensed protease inhibitors. We investigated the efficacy and tolerability of MK-5172
David Saadoun et al.
Annals of the rheumatic diseases, 73(5), 831-837 (2013-04-23)
The standard-of-care treatment of patients with hepatitis C virus (HCV)-mixed cryoglobulinemia (MC) vasculitis includes pegylated interferon α (PegIFN)-α plus ribavirin and/or rituximab. About 30-40% of patients are non-responders or relapsers to such combination. To analyse the safety and efficacy of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service