Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H97BAF1F8

Targetmol Chemicals Inc.

SEB Domain 144-153 acetate

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
1219.4

Iupac Name

L-lysyl-L-lysyl-L-lysyl-L-valyl-L-threonyl-L-alanyl-L-glutaminyl-L-glutamyl-L-leucyl-L-aspartic acid--acetic acid (1/1)

Inchi Code

1S/C50H90N14O17.C2H4O2/c1-25(2)23-34(47(77)62-35(50(80)81)24-38(69)70)61-45(75)33(17-19-37(67)68)60-44(74)32(16-18-36(55)66)57-41(71)27(5)56-49(79)40(28(6)65)64-48(78)39(26(3)4)63-46(76)31(15-9-12-22-53)59-43(73)30(14-8-11-21-52)58-42(72)29(54)13-7-10-20-51;1-2(3)4/h25-35,39-40,65H,7-24,51-54H2,1-6H3,(H2,55,66)(H,56,79)(H,57,71)(H,58,72)(H,59,73)(H,60,74)(H,61,75)(H,62,77)(H,63,76)(H,64,78)(H,67,68)(H,69,70)(H,80,81);1H3,(H,3,4)/t27-,28+,29-,30-,31-,32-,33-,34-,35-,39-,40-;/m0./s1

InChI key

KZCWIHYYZATKDS-JDABFKGFSA-N

storage temp.

-80/-20

Purity

99.22%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/SEB Domain 144-153 acetate(210229-94-0 free base)

MSDS

https://www.targetmol.com/compound/SEB Domain 144-153 acetate(210229-94-0 free base)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service