Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H97BAF133

Targetmol Chemicals Inc.

yibeissine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
443.63

Iupac Name

(3S,3'R,3a'S,4aS,6aS,6bS,6'S,7a'R,9R,11R,11aS,11bR)-3,11-dihydroxy-3',6',10,11b-tetramethyl-1,2,3,3a',4,4a,4',5',6a,6b,6',7,7',7a',8,11,11a,11b-octadecahydro-3'H-spiro[benzo[a]fluorene-9,2'-furo[3,2-b]pyridin]-5(6H)-one

Inchi Code

1S/C27H41NO4/c1-13-9-21-24(28-12-13)15(3)27(32-21)8-6-17-18-11-20(30)19-10-16(29)5-7-26(19,4)23(18)25(31)22(17)14(27)2/h13,15-19,21,23-25,28-29,31H,5-12H2,1-4H3/t13-,15+,16-,17-,18-,19+,21+,23+,24-,25-,26-,27-/m0/s1

InChI key

NURPXYQPDMVKOY-RUKZUWONSA-N

storage temp.

-80/-20

Purity

99.88%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/yibeissine

MSDS

https://www.targetmol.com/compound/yibeissine


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service