Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H97BAF0C4

Targetmol Chemicals Inc.

Bombesin acetate(31362-50-2 free base)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
1679.92

Iupac Name

N1-(11-((1H-imidazol-5-yl)methyl)-23-((1H-indol-3-yl)methyl)-43-amino-29-(2-amino-2-oxoethyl)-26-(3-amino-3-oxopropyl)-5-carbamoyl-43-imino-8,35-diisobutyl-17-isopropyl-20-methyl-7,10,13,16,19,22,25,28,31,34,37-undecaoxo-2-thia-6,9,12,15,18,21,24,27,30,33,36,42-dodecaazatritetracontan-38-yl)-2-(5-oxopyrrolidine-2-carboxamido)pentanediamide acetate

Inchi Code

1S/C71H110N24O18S.C2H4O2/c1-34(2)24-47(92-62(105)43(14-11-22-79-71(76)77)89-64(107)45(15-18-52(72)96)90-63(106)44-17-20-55(99)85-44)61(104)81-31-56(100)87-51(28-54(74)98)69(112)91-46(16-19-53(73)97)65(108)94-49(26-38-29-80-41-13-10-9-12-40(38)41)66(109)84-37(7)60(103)95-58(36(5)6)70(113)82-32-57(101)86-50(27-39-30-78-33-83-39)68(111)93-48(25-35(3)4)67(110)88-42(59(75)102)21-23-114-8;1-2(3)4/h9-10,12-13,29-30,33-37,42-51,58,80H,11,14-28,31-32H2,1-8H3,(H2,72,96)(H2,73,97)(H2,74,98)(H2,75,102)(H,78,83)(H,81,104)(H,82,113)(H,84,109)(H,85,99)(H,86,101)(H,87,100)(H,88,110)(H,89,107)(H,90,106)(H,91,112)(H,92,105)(H,93,111)(H,94,108)(H,95,103)(H4,76,77,79);1H3,(H,3,4)

InChI key

KWYGODXEILBTFB-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

98.44%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Bombesin acetate(31362-50-2 free base)

MSDS

https://www.targetmol.com/compound/Bombesin acetate(31362-50-2 free base)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service