Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z777127

SRT®-C SEC-2000 (5 µm) HPLC Columns

L × I.D. 5 cm × 7.8 mm, stainless steel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

SRT®-C SEC-2000 Gel Filtration Guard Column, 5 μm, L × I.D. 5 cm × 7.8 mm, 5 μm particle size, stainless steel

material

stainless steel

Agency

suitable for USP L59

product line

SRT®

mol wt

>10,000,000

manufacturer/tradename

Sepax 235980-7805

packaging

1 ea of

parameter

80 Mpa max. pressure (3000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

5 cm × 7.8 mm

impurities

<10 ppm metals

matrix

spherical silica
fully porous particle

matrix active group

hydrophilic polymer phase

particle size

5 μm

pore size

2000 Å

operating pH range

2.0-8.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

The bonded phase in SRT-C SEC columns is more polar than the hydrophilic phase used in SRT columns, making SRT-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies. SRT-C columns share with SRT columns the same 5 micron silica particle and the same wide range of pore sizes so you can select the pore size that will allow full access for your compound of interest.


SRT-C User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SRT is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service