Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G005790

LC/MS Analysis of L-Methionine Sulfoximine, Sulfone and Sulfoxide on Ascentis® Express HILIC

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express HILIC, 10 cm x 3.0 mm I.D., 2.7 μm particles (53970-U)
mobile phase: [A] acetonitrile; [B] water with 0.1% formic acid, pH 3.5 with ammonium formate; (75:25, A:B)
flow rate: 0.4 mL/min
pressure: 1350 psi 93 bar
temp.: 25 °C
detector: MS, ESI (+), TIC, m/z 150-300
injection: 5 μL
sample: 10 μg/mL in 90:10, methanol:water

suitability

application for HPLC

application(s)

life science and biopharma

Analysis Note

L-methionine sulfoximine and L-buthionine sulfoximine are used in are used to prevent additional enzyme activity in Chinese hampster ovary (CHO) cell lines with supplemental Glutamine Synthase (GS). It is of interest to be able to monitor both purity of such molecules in order to control addition to cell media as well as for the assay of parent molecules during use. The analytes are highly polar and thus should be amenable to HILIC-LC-MS analysis. This first installment investigates the ability of Ascentis Express HILIC and Ascentis Express OH5 stationary phases for the separation of the methionine analog and related compounds as well as methionine and buthionine separations.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    L-Methionine sulfone
    L-Methionine sulfoximine, suitable for cell culture
    L-Methionine sulfoxide

standard

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service