BOTB9089
Artemisia vulgaris Leaf
Botanical reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
BOTANICERT®
application(s)
food and beverages
pharmaceutical
format
matrix material
storage temp.
2-25°C
Related Categories
General description
English name: Mugwort, French name: Armoise commune
This botanical reference material (BRM) is manufactured by BotaniCERT™ and EXTRASYNTHESE™. The material is sourced from botanical gardens, collected by botanical experts during the flowering period, analyzed by seasoned phytochemists and geneticians.
BotaniCERT Botanical Reference Materials are samples ofplants or plant parts (leaf, root, rhizome, fruit, flower, seed...) authenticatedby organoleptic criteria, macroscopic and microscopic examination, HP-TLC andHPLC phytochemical fingerprint and genetic markers by PCR. They are given with2 or 3 markers (ITS, matK, rbcL) and DNA sequences are reported.
The products are provided as 5g samples and a detailed Certificate of Analysis, including related pictures, chromatograms and DNA sequences is available.
Using a Botanical Reference Material in an identification analysis allows calibration of methods and increases the guarantee of the result obtained.
This botanical reference material (BRM) is manufactured by BotaniCERT™ and EXTRASYNTHESE™. The material is sourced from botanical gardens, collected by botanical experts during the flowering period, analyzed by seasoned phytochemists and geneticians.
BotaniCERT Botanical Reference Materials are samples ofplants or plant parts (leaf, root, rhizome, fruit, flower, seed...) authenticatedby organoleptic criteria, macroscopic and microscopic examination, HP-TLC andHPLC phytochemical fingerprint and genetic markers by PCR. They are given with2 or 3 markers (ITS, matK, rbcL) and DNA sequences are reported.
The products are provided as 5g samples and a detailed Certificate of Analysis, including related pictures, chromatograms and DNA sequences is available.
Using a Botanical Reference Material in an identification analysis allows calibration of methods and increases the guarantee of the result obtained.
Application
For identity and authenticity testing of botanical materials
For more information, check out the on demand webinar "Design and Usage of Botanical Reference Materials"
For more information, check out the on demand webinar "Design and Usage of Botanical Reference Materials"
Legal Information
BOTANICERT is a registered trademark of BOTANICERT, Société, FR
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service