Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

796-0150

HPLC Analysis of Acidic Herbicides in Water on a Polymeric C18 Column after SPE using Supelclean ENVI-Carb

application for SPE, application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable
solid phase extraction (SPE): suitable

test parameters

sample preparation: SPE (Solid Phase Extraction)
sample/matrix: Fresh 1 L water samples, dechlorinated with sodium thiosulfate when necessary, at ambient temperature and pH.
SPE tube/cartridge: Supelclean ENVI-Carb, 250 mg/6 mL (57092)
condition: 10 mL DI water at 20 mL/min
sample addition: 0.9 L water sample at 20 mL/min
drying: 10 min using clean nitrogen
washing: 10 mL DI water at 20 mL/min
elution: 10 mL 0.1% phosphoric acid in methylene chloride:acetonitrile (80:20) at 5 mL/min
column: polymeric-coated silica-based PAH specialty column, 20 cm × 3 mm I.D., 5 μm (Supelco equivalent, SUPELCOSIL LC-PAH, available upon request)
mobile phase: gradient, [A]: 0.05% phosphoric acid in DI water; [B]: acetonitrile
flow rate: 0.5 mL/min
column temp.: 50 °C
detector: photodiode array- peak width: 0.053 min, sampling interval: 0.320 sec, monitor 210 nm & 225 nm
injection: 10 μL of extract (4-5 ppb each analyte in water)

suitability

application for HPLC
application for SPE

application(s)

environmental

Application

using Zymark AutoTrace Extraction WorkStation 1.20

Legal Information

ENVI is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    Dinoseb
    2,4-D, analytical standard
    Picloram, BioReagent, suitable for plant cell culture
    2,4,5-T, PESTANAL®
    Pentachlorophenol
    Tetrachloroterephthalic acid
    Dicamba

SPE tube or plate

analytical column


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service