Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

503126

Supelco

Supelpure-O Oxygen/Moisture Trap

S-trap, 1/8 in. fitting

Synonym(s):

S-TRAP, SUPELPURE-O, 1/8IN FITTINGS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

fitting

1/8 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

The oxygen-removing catalysts can reduce oxygen to less than 2 ppb when the level in the incoming gas does not exceed 10 ppm. Because the catalyst is coated on a Molecular Sieve, this trap also can remove water vapor.

Dimensions:
  • 120 cc Trap: 111/8in. (28.2 cm) long x 13/8 in. (35 mm) diameter
  • 750 cc Trap: 161/2 in. (41.9 cm) long x 25/16in. (59 mm) diameter
  • S-Trap: 71/2 in. (19 cm) long x 59/16 in. (14.1 cm) wide x 1/2 in. (13 mm) diameter [total bed length = 193/4 in. (50.2 cm); the extended bed length ensures prolonged contact between the gas and the adsorbent and provides greater working capacity]

Application

It has been used in purifying ethylene during slurry polymerization for preparing polymer with medium molecular weight distributions; in a study performed to understand fragmentable support of organic nanoparticles for Ziegler–Natta Catalysts.

Legal Information

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic nanoparticles as fragmentable support for Ziegler?Natta catalysts.
Nietzel, Sven, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 53.1, 15-22 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service