Skip to Content
MilliporeSigma
All Photos(1)

Documents

49155

Supelco

Polynuclear Aromatic Hydrocarbons Mix

certified reference material, 500 μg/mL each component in toluene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

500 μg/mL each component in toluene

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

Polynuclear aromatic hydrocarbons is a group of environmental organic micro-pollutants, which can be released into the environment, due to various factors like the incomplete combustion of organic materials and compounds, fossil fuels, forest fires and also microbiological synthesis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    Anthracene
    Benz[a]anthracene
    Benzo[a]pyrene
    Benzo[e]pyrene
    Chrysene
    Fluoranthene
    Perylene
    Phenanthrene
    Pyrene
    Triphenylene
    See All (10)

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

45.0 °F

Flash Point(C)

7.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perovskites and Related Mixed Oxides: Concepts and Applications, Volume 1, 1 (2016)
Comparison of different drying, extraction and detection techniques for the determination of priority polycyclic aromatic hydrocarbons in background contaminated soil samples
Berset.D.J, et al.
Analytica Chimica Acta, 383, 263-275 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service