Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47798

Supelco

4-Chlorotoluene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4Cl
CAS Number:
Molecular Weight:
126.58
Beilstein:
1903635
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

4.38 (vs air)

vapor pressure

10 mmHg ( 45 °C)

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.52 (lit.)

bp

162 °C (lit.)

mp

6-8 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

Cc1ccc(Cl)cc1

InChI

1S/C7H7Cl/c1-6-2-4-7(8)5-3-6/h2-5H,1H3

InChI key

NPDACUSDTOMAMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Zewdie et al.
Journal of toxicology and environmental health, 50(2), 159-172 (1997-02-07)
Treatment of rats with p-chlorotoluene (PCT, 1 g/kg, ip) resulted in peak PCT blood and lung concentrations at 4 h, which declined to very low levels at 12 h. The concentration of PCT in the liver attained its highest value
C Arunagiri et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1747-1756 (2011-06-18)
The FT-IR and FT-Raman spectra of 2-bromo-4-chlorotoluene (2B4CT) molecule have been recorded in the region 4000-400 cm(-1) and 3500-50 cm(-1), respectively. Optimized geometrical structures, harmonic vibrational frequencies, intensities, reduced mass, force constants and depolarization ratio have been computed by the
T Nishio et al.
Applied microbiology and biotechnology, 55(3), 321-325 (2001-05-09)
p-Cymene monooxygenase (CMO) from Pseudomonas putida F1 consists of a hydroxylase (CymA1) and a reductase component (CymA2) which initiate pcymene (p-isopropyltoluene) catabolism by oxidation of the methyl group to p-isopropylbenzyl alcohol (p-cumic alcohol). To study the possible diverse range of
Ya-Jun Liu et al.
Physical chemistry chemical physics : PCCP, 7(23), 3938-3942 (2005-12-07)
Equilibrium geometries and vibrational frequencies of the ground and some excited states of p-chlorotoluene were calculated by the complete active space self-consistent field (CASSCF) method. Multi-reference CASSCF second order perturbation theory (MSCASPT2) calculations were performed on the vertical excitation energies
Mannar R Maurya et al.
Dalton transactions (Cambridge, England : 2003), (32)(32), 4220-4232 (2008-08-07)
3-Formylsalicylic acid (Hfsal), covalently bound to chloromethylated polystyrene (PS) and cross-linked with 5% divinylbenzene reacts with d,l-alanine and l-isoleucine to give the Schiff-base tridentate ligands PS-H(2)fsal-d,l-Ala and PS-H(2)fsal-l-Ile, respectively. These anchored ligands upon reaction with VOSO(4) and Cu(CH(3)COO)(2).H(2)O form the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service