Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442283-U

Supelco

2,3,4-Trichlorophenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OH
CAS Number:
Molecular Weight:
197.45
Beilstein:
1945113
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

75-79 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Oc1ccc(Cl)c(Cl)c1Cl

InChI

1S/C6H3Cl3O/c7-3-1-2-4(10)6(9)5(3)8/h1-2,10H

InChI key

HSQFVBWFPBKHEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanjun Fang et al.
Journal of proteomics, 110, 172-182 (2014-08-12)
Pentachlorophenol (PCP) and 2,4,6-trichlorophenol (TCP) are suspected of disrupting the endocrine system and thus affecting human and wildlife reproduction, but the potential common mechanisms and biomarkers of chlorophenols (CPs) in the ovary are not fully elucidated. In the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service