33272-U
Crimpers
Hand crimper, for use with 8mm seals, pkg of 1 ea
Synonym(s):
HAND CRIMPER 8MM
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 1 ea
compatibility
for use with 8mm seals
Looking for similar products? Visit Product Comparison Guide
General description
The Supelco hand held crimper provides a consistent and dependable seal that allows secure vial closure each and evey time. The high quality construction is engineered for durability and long life to provide a smooth and simple operation.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service