Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB2476

Sigma-Aldrich

Anti-GRB2 Antibody, clone 1J2 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Adapter protein GRB2, Growth factor receptor-bound protein 2, Protein Ash, SH2/SH3 adapter GRB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... GRB2(9402)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1J2 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Growth factor receptor-bound protein 2 (GRB2). It targets an epitope within 15 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal half of human Growth factor receptor-bound protein 2 (GRB2).

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected GRB2 in HEK293 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected GRB2 in HeLa, PC12, and RAW264.7 cell lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected GRB2 in HEK293 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected GRB2 in Human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Growth factor receptor-bound protein 2 (UniProt: P62993; also known as Adapter protein GRB2, Protein Ash, SH2/SH3 adapter GRB2) is encoded by the GRB2 (also known as ASH) gene (Gene ID: 2885) in human. GRB2 is a signaling adaptor that is ubiquitously expressed and interacts with numerous receptors and intracellular signaling molecules. It contains one central SH2 domain flanked by two SH3 domains and is broadly expressed in many tissues and is shown to be essential for embryonic development and multiple cellular functions. It is a major mediator in Ras-MAPK activation pathway induced by numerous receptors because of its association with son of sevenless (SoS). GRB2 plays a role in early T cell development  and is shown to be a positive regulator of TCR signaling. It does not directly affect the activities of Ras and Erk1/2; however, it amplifies the activity of Lck, a tyrosine kinase that functions at the top of the TCR signaling cascade. It exists in a constitutive equilibrium between monomeric and dimeric states. The monomeric form is capable of binding to SoS and upregulating MAP kinase signaling, and the dimeric state is inhibitory to this process. Phosphorylation of tyrosine 160 or binding of a tyrosylphosphate-containing ligand to the SH2 domain of Grb2, results in dimer dissociation. Phosphorylation of tyrosine 160 is readily detectable in the malignant prostate, colon. and breast cancers.  Two isoforms of GRB2 have been described that are produced by alternative splicing. Isoform 2 acts as a dominant negative protein that suppresses proliferative signals, may trigger active programmed cell death. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Ahmed, Z., et al. (2015). Nat. Commun. 6; Article7354; Jang, IK., et al. (2011). Proc. Natl. Acad. Sci. USA. 108 (19); 7926-7931; Fath, I., et al. (1994). Science. 264(5161); 971-974; Lowenstein, EJ., et al. (1992). Cell. 70(3); 431-442).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service