Skip to Content
MilliporeSigma
All Photos(11)

Key Documents

ZRB2045

Sigma-Aldrich

Anti-UBC9/UBE2I Antibody, clone 2H5 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

SUMO-conjugating enzyme UBC9;RING-type E3 SUMO transferase UBC9;SUMO-protein ligase;Ubiquitin carrier protein 9;Ubiquitin carrier protein I;Ubiquitin-conjugating enzyme E2 I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2H5, recombinant monoclonal

description

2H5 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 18.01 kDa
observed mol wt ~18 kDa

purified by

using Protein A

species reactivity

monkey, human, mouse

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... UBE2I(7329)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2H5 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detect UBC9/UBE2I.

Immunogen

His-tagged recombinant fragment corresponding to 145 amino acids ( aa 2-146) of human UBC9.

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected UBC9/UBE2I in Jurkat cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected UBC9/UBE2I in U251 MG, NIH3T3, and COS-7 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected UBC9/UBE2I in human fallopian tube tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound UBC9/UBE2I recombinant fragment with a KD of 4.0 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected UBC9/UBE2I in Jurkat cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

SUMO-conjugating enzyme UBC9 (UniProt: P63279; also known as RING-type E3 SUMO transferase UBC9, SUMO-protein ligase, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, Ubiquitin-conjugating enzyme E2 I) is encoded by the UBE2I (also known as UBC9, UBCE9) gene (Gene ID: 7329) in human. SUMOylation is process that adds a small ubiquitin-like modifier (SUMO) polypeptide to the -amino group of a lysine residue of proteins. UBC9 is a SUMO-conjugating enzyme 2 ligase that is expressed in multiple tissues, including heart, skeletal muscle, pancreas, kidney, liver, lung, placenta, brain, testis, and thymus. It accepts SUMO1, SUMO2, SUMO3, SUMO4 and SUMO1P1/SUMO5 from the UBLE1A-UBLE1B E1 complex and catalyzes their covalent attachment to other proteins with the help of an E3 ligase. It can catalyze the formation of poly-SUMO chains. Its UBC domain is localized in amino acids 4-157. It is reported to sumoylate p53/TP53 at lysine 386. Its expression is shown to increase during reprogramming of mouse embryonic fibroblasts (MEFs) into induced pluripotent stem (iPS) cells and its suppression inhibits iPS cell induction. Knockdown of UBC9 is reported to decreases the expression of Nanog, Klf4, Oct4, and Sox2 in embryonic stem cells. UBC9 also has a cardioprotective role and is shown to improve cardiac function and ease left ventricle remodeling following myocardial infarction. Phosphorylation of serine 71 of UBC9 enhances its SUMOylation activity. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Xiao, Q., et al. (2020). Front. Pharmacol. 11; 561306; Tahmasebi, S., et al. (2014). Stem Cells. 32(4); 1012-1020; Gupta, MK., et al. (2014). Cir. Res. 115(8); 721-729).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

0.3 mg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service