Skip to Content
MilliporeSigma
All Photos(9)

Documents

ZRB1662

Sigma-Aldrich

Anti-p-RPA2-Ser4/8 Antibody, clone 2F23 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Replication protein A 32 kDa subunit;RP-A p32;Replication factor A protein 2;RF-A protein 2;Replication protein A 34 kDa subunit;RP-A p34

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2F23, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 29.25 kDa
observed mol wt ~35 and 38 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer4/8)

Gene Information

human ... RPA2(6118)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2F23 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Replication protein A 32 kDa subunit (RPA2) phosphorylated on serine 4 and 8.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphoserines 4 and 8 from the N-terminal region of human Replication protein A 32 kDa subunit (RPA2).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HeLa cells treated with Etoposide.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected RPA2 phosphorylated on serine 4 and 8 in lysate from HeLa cells, overnight serum starved and treated with Etoposide ( 20 g/mL; 3 h)

Tested applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Phospho-RPA2-Ser4/8 in human lung cancer tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Phospho-RPA32-Ser4/8 peptide with a KD of 2.7 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in lysate from HeLa cells treated with Etoposide.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected RPA2 phosphorylated on serine 4 and 8 in lysate from HeLa cells, overnight serum starved and treated with Etoposide ( 20 g/mL; 3 h).

Target description

Replication protein A 32 kDa subunit (UniProt: P15927; also known as RP-A p32, Replication factor A protein 2, RPA2, RF-A protein 2, Replication protein A 34 kDa subunit, RP-A p34) is encoded by the RPA2 (also known as REPA2, RPA32, RPA34) gene (Gene ID: 6118) in human. RPA2 is a part of the heterotrimeric replication protein A complex (RPA/RP-A) that binds and stabilizes single-stranded DNA intermediates that form during DNA replication or upon DNA stress. It prevents their reannealing and in parallel, recruits and activates different proteins and complexes involved in DNA metabolism. It also plays an essential role in DNA replication. It is translationally up-regulated in response to DNA damage. In the cellular response to DNA damage, the RPA complex controls DNA repair and DNA damage checkpoint activation. Through recruitment of ATR interacting protein (ATRIP), it activates the ATR kinase. It is also required for the recruitment of the DNA double-strand break repair factors RAD51 and RAD52 to chromatin in response to DNA damage. It also recruits XPA and XPG that are involved in nucleotide excision repair. RPA2 is differentially phosphorylated throughout the cell cycle, becoming phosphorylated at the G1-S transition, and dephosphorylated in late mitosis. It is mainly phosphorylated at serine 23 and 29 by cyclin A-CDK2 and cyclin B-CDK1, respectively. It becomes hyperphosphorylated on additional residues including serine 4 and 8, threonine 21, and serine 33 in response to DNA damage. This hyperphosphorylation is mediated by ATM, ATR, DNA-PK. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Yin, J-Y., et al. (2013). Carcinogenesis. 34(6); 1224-1231; Sleeth, KM., et al. (2007). J. Mol. Biol. 373(1); 38-47; Erdile, LF., et al. (1990). J. Biol. Chem. 265(6); 3177-3182).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service