Documents
Z659436
Bandelin SONOPULS ultrasonic homogenizer accessories
Indirect processing cup booster BR 30
Synonym(s):
high-intensity sonication vessel
About This Item
UNSPSC Code:
41101520
NACRES:
NB.74
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 7510
availability
available only in EU
General description
For high-intensive irradiation of smallest and sensitive sample volumes, e. g. cleaning of radioactively contaminated seeds, cell disruption or treatment of pathogens and toxic material. No cross-contamination or sample loss. Flow-through cooling liquid avoids rise of temperature