Key Documents
Z358460
Sigma-Aldrich
Carestream® BioMax® MR film
Synonym(s):
autoradiography film, xray film
About This Item
UNSPSC Code:
41105341
NACRES:
NB.22
size
9.5 in. (24 cm) × 12 in. (30 cm)
General description
Sheets of film with no interleaved paper
Single-emulsion film that provides maximum resolution and clearest background. The most sensitive film for direct 14C, 35S, and 33P autoradiography. Higher resolution than X-OMAT AR and BioMax MS for 32P direct and screen exposures and chemiluminescence, but with reduced sensitivity.
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Shujuan Gao et al.
Journal of molecular signaling, 6, 8-8 (2011-08-13)
The family of A-kinase-anchoring proteins, AKAPs, constitutes a group of molecular scaffolds that act to catalyze dynamic interactions of protein kinase A, protein kinase C, tyrosine kinases, G-protein-coupled receptors and ion channels. AKAP5 (MW ~47 kDa) and AKAP12 (MW ~191
Anna-Lena Ruppert et al.
Journal of molecular histology, 51(4), 421-435 (2020-07-04)
Tuft cells are a rare population of chemosensory cells at the mucosal surface epithelia of hollow organs. Their name-giving morphological feature is an apical tuft of stiff microvilli. Accordingly, the actin-binding protein, villin, was identified as one of the first
Atsuko Kimura et al.
The European journal of neuroscience, 30(2), 299-306 (2009-07-21)
The 5-HT(2C) receptor has been implicated in mood and eating disorders. In general, it is accepted that 5-HT(2C) receptor agonists increase anxiety behaviours and induce hypophagia. However, pharmacological analysis of the roles of these receptors is hampered by the lack