Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WH0023395M3

Sigma-Aldrich

Monoclonal Anti-LARS2 antibody produced in mouse

clone 3F12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-KIAA0028, Anti-LEURS, Anti-MGC26121, Anti-leucyl-tRNA synthetase 2, mitochondrial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LARS2(23395)

General description

This gene encodes a class 1 aminoacyl-tRNA synthetase, mitochondrial leucyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. (provided by RefSeq)

Immunogen

LARS2 (NP_056155, 806 a.a. ~ 903 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VPRKLCAHYTWDASVLLQAWPAVDPEFLQQPEVVQMAVLINNKACGKIPVPQQVARDQDKVHEFVLQSELGVRLLQGRSIKKSFLSPRTALINFLVQD

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service