Key Documents
TMO374500-BR
Nunc® Universal Tubes in 2D bar coded latch rack
polypropylene, capacity (1.8 mL), flat bottom, external thread, untreated, case of 10 × 24 ea
Synonym(s):
bar coded tube storage, traceable tube storage
About This Item
UNSPSC Code:
41122800
NACRES:
NB.11
material
flat bottom
polypropylene
packaging
case of 10 × 24 ea
manufacturer/tradename
Nunc 374500-BR
capacity
(1.8 mL)
General description
Experience the best features of the Thermo Scientific storage tube portfolio: universal cap, universal latch rack, and free-standing tube. This versatile 2.0mL internally-threaded tube is ideal both for a large storage system like a biobank, and in smaller scale for traditional cryostorage. **Now available in a barcoded rack to allow tracking of the rack in addition to individual tubes. Sample identity is ensured due to a permanently attached 2D barcode on each tube and permanent laser etching of the rack. All tubes and racks are supplied free from DNA, RNase, DNase, and endotoxins.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries