Skip to Content
MilliporeSigma
All Photos(1)

Documents

T3407

Sigma-Aldrich

Terpineol

~95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

description

mixture of isomers

Assay

~95%

composition

α-terpineol, ~65%
β-terpineol, ~10%
γ-terpineol, ~20%

bp

213-218 °C (lit.)

density

0.934 g/mL at 20 °C (lit.)
0.93 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C1CCC(C)(O)CC1.CC2=CCC(CC2)C(C)(C)O

InChI

1S/2C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11;1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;9,11H,1,4-7H2,2-3H3

InChI key

MGABGZGUMNDCSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng-Chang Wang et al.
Sensors (Basel, Switzerland), 15(6), 14286-14297 (2015-06-20)
Highly sensitive H2 gas sensors were prepared using pure and Pt-loaded SnO2 nanoparticles. Thick film sensors (~35 μm) were fabricated that showed a highly porous interconnected structure made of high density small grained nanoparticles. Using Pt as catalyst improved sensor
Khaled Sebei et al.
Biological research, 48, 7-7 (2015-02-06)
In this paper, we have studied the essential oils chemical composition of the leaves of seven Eucalyptus species developed in Tunisia. Eucalyptus leaves were picked from trees growing in different arboretums in Tunisia. Choucha and Mrifeg arboretums located in Sedjnene
Yi-Fei Sun et al.
Nanoscale, 7(25), 11173-11181 (2015-06-11)
To date, most investigations of Ni-Fe bimetallic catalysts for solid oxide fuel cells (SOFCs) have focused on materials with micro-scale particle sizes, which severely restrict their catalytic activity. In this study, we fabricated a Ni- and/or Fe-doped A-site-deficient LaSrCrO3 perovskite
Dae-Yong Son et al.
Physical chemistry chemical physics : PCCP, 17(23), 15193-15200 (2015-05-21)
To understand the role of the dye/oxide interface, a model system using a nanocrystalline SnO2 and 3-hexyl thiophene based MK-2 dye is proposed. A thin interfacial TiO2 blocking layer (IBL) is introduced in between SnO2 and MK-2 and its effects
Han Fu et al.
Nanoscale research letters, 9(1), 631-631 (2014-12-19)
Fabricating functional compounds on substrates with complicated morphology has been an important topic in material science and technology, which remains a challenging issue to simultaneously achieve a high growth rate for a complex nanostructure with simple controlling factors. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service