Skip to Content
MilliporeSigma
All Photos(1)

Documents

T2825

Sigma-Aldrich

TIQ-A

≥98% (HPLC), solid

Synonym(s):

Thieno[2,3-c]isoquinolin-5-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7NOS
CAS Number:
Molecular Weight:
201.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: soluble 16 mg/mL

storage temp.

2-8°C

SMILES string

O=C1Nc2sccc2-c3ccccc13

InChI

1S/C11H7NOS/c13-10-8-4-2-1-3-7(8)9-5-6-14-11(9)12-10/h1-6H,(H,12,13)

InChI key

LQJVOLSLAFIXSV-UHFFFAOYSA-N

Application

TIQ-A has been used as a PAPR-1 inhibitor to study eosinophilic infiltration into the airways of OVA-challenged mice1. It has also been used to study its role in the regression of atherosclerotic plaques in high-fat diet-fed ApoE(-/-) mice2.

Biochem/physiol Actions

TIQ-A is a potent inhibitor of PARP, anti-apoptotic.

Preparation Note

TIQ-A is soluble in DMSO at 16 mg/ml.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chetan P Hans et al.
The Journal of pharmacology and experimental therapeutics, 329(1), 150-158 (2009-01-07)
We recently showed that poly(ADP-ribose) polymerase (PARP) is activated within atherosclerotic plaques in an animal model of atherosclerosis. Pharmacological inhibition of PARP or reduced expression in heterozygous animals interferes with atherogenesis and may promote factors of plaque stability, possibly reflecting
Mustapha Oumouna et al.
Journal of immunology (Baltimore, Md. : 1950), 177(9), 6489-6496 (2006-10-24)
We recently used a murine model of allergic airway inflammation to show that poly(ADP-ribose) polymerase-1 (PARP-1) plays an important role in the pathogenesis of asthma-related lung inflammation. In this study, we show that PARP-1 inhibition, by a novel inhibitor (TIQ-A)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service