Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T165

Sigma-Aldrich

S(+)-Terguride

solid

Synonym(s):

S(+)-N,N-Diethyl-N′([8α]-6-methylergolin-8-yl)urea, S(+)-Terguride (5R, 8S, 10R)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N4O
CAS Number:
Molecular Weight:
340.46
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

optical activity

[α]20/D +28°, c = 0.2 in pyridine(lit.)

color

white

solubility

DMSO: soluble
H2O: insoluble
ethanol: soluble

SMILES string

[H][C@@]12Cc3c[nH]c4cccc(c34)[C@@]1([H])C[C@@H](CN2C)NC(=O)N(CC)CC

Gene Information

human ... DRD2(1813)
rat ... Drd2(24318)

Biochem/physiol Actions

Ergot derivative; dehydrogenated analog of lisuride; D2 dopamine receptor partial agonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Akai et al.
Annals of neurology, 33(5), 507-511 (1993-05-01)
Behavioral effects of terguride, a partial dopamine D2 agonist, on 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-lesioned parkinsonian cynomolgus monkeys were compared with those of the dopamine agonist apomorphine and the dopamine antagonist haloperidol. Terguride alone ameliorated the parkinsonism without inducing any sign of excitability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service