Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1442

Eppendorf® Thermomixer Compact

with 1.5 mL block, AC/DC input 220 V AC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

manufacturer/tradename

Eppendorf® 022670051 (USA)
Eppendorf® 5350 000.013 (European)
Eppendorf® 5350 000.030 (UK)

W × D × H

20.6 cm × 30.4 cm × 17.0 cm

Looking for similar products? Visit Product Comparison Guide

General description

Incubation and mixing may be performed independently or simultaneously. Includes autoclavable and freezer-safe numbered rack with 24 numbered positions make processing and transporting samples quick and easy. Temperature Control Range - 4°C above ambient up to 99°C in 1°C increments.
This new thermomixer combines heating and shaking of 24 × 1.5 mL samples at a time to ease user workload. Incubation and mixing may also be performed independently. Includes autoclavable and freezer-safe numbered rack with 24 numbered positions make processing and transporting samples quick and easy.

  • Short-mix function for quick vortex applications
  • Mixing speed - 300 to 1,300 rpm
  • Mixing stroke - 3 mm
  • Temperature Control Range: 4°C above ambient up to 99°C in 1°C increments
  • Temperature Accuracy - ± 0.5°C at 20 to 45 °C

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengfei Xu et al.
Toxicology letters, 290, 29-35 (2018-03-20)
Constitutive androstane receptor (CAR) is a nuclear receptor that not only regulates drug-metabolizing enzymes but also influences energy metabolism. TC, 1, 4-bis [2-(3, 5-dichloropyridyloxy)] benzene (TCPOBOP) has been shown to inhibit lipogenesis in the liver and adipose tissues. The mammary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service