Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6212

Sigma-Aldrich

sDLL-1 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

Delta-1, Delta-like protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

57-60 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DLL-1(28514)

General description

Human sDLL-1 comprises the extracellular signaling domain of DLL-1, a member of a structurally-related family of single-pass type I transmembrane proteins that serve as ligands for Notch receptors. It is expressed in the heart and pancreas, and to a lesser extent in various other tissues. Recombinant human sDLL-1 is a 57.0-60.0 kDa glycoprotein containing 522 amino-acid residues. The DLL-1 gene is mapped to human chromosome 6q27.

Biochem/physiol Actions

DLL-1 (delta-like ligand 1) functions to specifically activate the Notch-1 and Notch-2 receptors. The Notch signaling pathway regulates endothelial-cell differentiation, proliferation and apoptosis, and is essential for the development, maintenance and remodeling of the vascular system. DLL-1 suppresses differentiation of hematopoietic progenitor cells into the B-cell lineage while promoting differentiation to T-cell and NK (natural killer) cell precursors. Upregulation of DLL-1 was observed in choriocarcinoma and hepatocellular carcinoma.

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized from 1X PBS, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notch receptor?ligand binding and activation: Insights from molecular studies.
Chillakuri C R, et al.
Seminars in Cell & Developmental Biology (2012)
Fine mapping of genes within the IDDM8 region in rheumatoid arthritis.
Hinks A, et al.
Arthritis Research & Therapy, 8(5), R145-R145 (2006)
Molecular genetics of T cell development.
Rothenberg E V and Taghon T
Annual Review of Immunology, 23 (2005)
Notch ligand Delta-like 1 as a novel molecular target in childhood neuroblastoma.
Bettinsoli P, et al.
BMC Cancer, 17(1), 352-352 (2017)
Notch receptor?ligand binding and activation: Insights from molecular studies.
Chillakuri C R, et al.
Seminars in Cell & Developmental Biology, 23(4), 421-428 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service