Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP4869

Sigma-Aldrich

Placental Lactogen human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

CSA, CSH1, CSH2, CSMT, FLJ75407, PL

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~22.3 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell based assay: suitable

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CSH1(1442)

General description

Human placental lactogen (HPL), also called human chorionic somatomammotropin 1, is a polypeptide placental hormone. Its structure and function is similar to that of growth hormone. The gene encoding this protein is localized on human chromosome 17. Recombinant human placental lactogen is a 22.3kDa protein.

Biochem/physiol Actions

Human placental lactogen (HPL) modifies the metabolic state of the mother during pregnancy to facilitate the energy supply, glucose supply and growth of the fetus. Studies show that targeted expression of this protein in transgenic mice lead to an increase in proliferation of β-cells.

Physical form

Sterile filtered and lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zinuo Chen et al.
Cell & bioscience, 11(1), 45-45 (2021-03-01)
In the urgent campaign to develop therapeutics against SARS-CoV-2, natural products have been an important source of new lead compounds. We herein identified two natural products, ginkgolic acid and anacardic acid, as inhibitors using a high-throughput screen targeting the SARS-CoV-2
A study of association between common variation in the growth hormone-chorionic somatomammotropin hormone gene cluster and adult fasting insulin in a UK Caucasian population.
Freathy RM
Journal of Negative Results in Biomedicine, 5, 18-18 (2006)
Meltem Muftuoglu et al.
Nucleic acids research, 34(1), 295-304 (2006-01-18)
Cockayne syndrome (CS) is a rare inherited human genetic disorder characterized by UV sensitivity, severe neurological abnormalities and prageroid symptoms. The CS complementation group B (CSB) protein is involved in UV-induced transcription coupled repair (TCR), base excision repair and general
Placental lactogen is expressed but is not translated into protein in breast cancer.
Tuttle TR
PLoS ONE, 9(1), e87325-e87325 (2014)
Jaeyong Cho et al.
Biochemical and biophysical research communications, 579, 110-115 (2021-10-02)
The liver increases its size during pregnancy to adapt to metabolic demand associated with pregnancy. Our previous study showed that proliferation of maternal hepatocytes are increased during pregnancy in mice and that estradiol (E2) is one of the candidate hormones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service