Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3147

Sigma-Aldrich

GSK046

≥98% (HPLC)

Synonym(s):

iBET-BD2, 4-acetamido-3-fluoro-N-(trans-4-hydroxycyclohexyl)-5-[(1S)-1-phenylethoxy]benzamide; 4-Acetamido-3-fluoro-N-((1R,4S)-4-hydroxycyclohexyl)-5-((S)-1-phenylethoxy)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27FN2O4
CAS Number:
Molecular Weight:
414.47
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

2-8°C

SMILES string

O=C(N[C@H]1CC[C@H](O)CC1)C2=CC(F)=C(NC(C)=O)C(O[C@H](C3=CC=CC=C3)C)=C2

General description

GSK046 is an orally bioavailable small molecule. Its structure contains a poly-substituted phenyl ring in the center. GSK046 reduces immuno-inflammation by inhibiting the production of monocyte chemoattractant protein 1 (MCP-1), a chemokine involved in the regulation of macrophage activation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Progress in the development of domain selective inhibitors of the bromo and extra terminal domain family (BET) proteins
Fu Y, et al.
European Journal of Medicinal Chemistry (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service