Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1891

Sigma-Aldrich

PK11007

≥98% (HPLC)

Synonym(s):

5-Chloro-2-(methylsulfonyl)-4-pyrimidinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5ClN2O4S
CAS Number:
Molecular Weight:
236.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C6H5ClN2O4S/c1-14(12,13)6-8-2-3(7)4(9-6)5(10)11/h2H,1H3,(H,10,11)

InChI key

WZUPWJVRWIVWEF-UHFFFAOYSA-N

Biochem/physiol Actions

PK11007 is a thiol-reactive anticancer agent that stabilizes wild type and mutant p53 through selective alkylation of two surface cysteines without interfering with its DNA binding. Additionally independently from p53, PK11007 impairs reactive oxygen species (ROS) detoxification in a number of mutant p53 cancer cells. PK11007 exhibits anticancer activity in number of cancer cell lines while shoving low cytotoxicity toward normal cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias R Bauer et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(36), E5271-E5280 (2016-08-24)
The tumor suppressor p53 has the most frequently mutated gene in human cancers. Many of p53's oncogenic mutants are just destabilized and rapidly aggregate, and are targets for stabilization by drugs. We found certain 2-sulfonylpyrimidines, including one named PK11007, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service