Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB5702988

Sigma-Aldrich

Anti-RNF20 Antibody, clone 8N4O8, Rabbit Monoclonal

Synonym(s):

BRE1, BRE1A, hBRE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

8N4O8, monoclonal

form

liquid

mol wt

120 kDa

species reactivity

mouse, mouse

concentration

0.65 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MSGIGNKRAAGEPGTSMPPEKKAAVEDSGTTVETIKLGGVSSTEELDIRTLQTKNRKLAEMLDQRQAIEDELREHIEKLERRQATDDASLLIVNRYWSQF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF20(56254)

General description

The protein encoded by this gene shares similarity with BRE1 of S. cerevisiae. The protein encoded by this human gene is an E3 ubiquitin ligase that regulates chromosome structure by monoubiquitinating histone H2B. This protein acts as a putative tumor suppressor and positively regulates the p53 tumor suppressor as well as numerous histone H2A and H2B genes. In contrast, this protein also suppresses the expression of several protooncogenes and growth-related genes, including many genes that are induced by epidermal growth factor. This gene selectively suppresses the expression of some genes by interfering with chromatin recruitment of transcription elongation factor SII (TFIIS). [provided by RefSeq, Feb 2012]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RNF20 (Q5VTR2).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service