Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5702462

Sigma-Aldrich

Anti-Integrin alpha 4 (ITGA4/CD49d) Antibody, clone 8M8C2, Rabbit Monoclonal

Synonym(s):

CD49D, IA4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

material

colorless

clone

8M8C2, monoclonal

form

liquid

species reactivity

human, rat, rat

concentration

0.60 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
immunoprecipitation (IP): 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

ALKFEIRATGFPEPNPRVIELNKDENVAHVLLEGLHHQRPKRYFTIVIISSSLLLGLIVLLLISYVMWKAGFFKRQYKSILQEENRRDSWSYINSKSNDD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGA4(3676)

General description

The gene encodes a member of the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain that function in cell surface adhesion and signaling. The encoded preproprotein is proteolytically processed to generate light and heavy chains that comprise the alpha 4 subunit. This subunit associates with a beta 1 or beta 7 subunit to form an integrin that may play a role in cell motility and migration. This integrin is a therapeutic target for the treatment of multiple sclerosis, Crohn's disease and inflammatory bowel disease. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Oct 2015]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 933-1032 of human Integrin alpha 4 (ITGA4/CD49d) (P13612).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC, IP

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service