Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5702444

Sigma-Aldrich

Anti-Rad50 Antibody, clone 5M2K10, Rabbit Monoclonal

Synonym(s):

NBSLD, RAD502, hRad50

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

5M2K10, monoclonal

form

liquid

mol wt

154 kDa

species reactivity

rat, human, rat

concentration

0.85 mg/mL

technique(s)

immunoprecipitation (IP): 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

RCSAGQKVLASLIIRLALAETFCLNCGIIALDEPTTNLDRENIESLAHALVEIIKSRSQQRNFQLLVITHDEDFVELLGRSEYVEKFYRIKKNIDQCSEIV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD50(10111)

General description

The protein encoded by this gene is highly similar to Saccharomyces cerevisiae Rad50, a protein involved in DNA double-strand break repair. This protein forms a complex with MRE11 and NBS1. The protein complex binds to DNA and displays numerous enzymatic activities that are required for nonhomologous joining of DNA ends. This protein, cooperating with its partners, is important for DNA double-strand break repair, cell cycle checkpoint activation, telomere maintenance, and meiotic recombination. Knockout studies of the mouse homolog suggest this gene is essential for cell growth and viability. Mutations in this gene are the cause of Nijmegen breakage syndrome-like disorder.[provided by RefSeq, Apr 2010]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1200-1300 of human Rad50 (Q92878).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB,IP

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service