Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1411646

Sigma-Aldrich

Anti-CER1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DAND4, MGC119894, MGC119895, MGC96951

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 30.1 kDa

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CER1(9350)

General description

This gene encodes a cytokine member of the cysteine knot superfamily, characterized by nine conserved cysteines and a cysteine knot region. The cerberus-related cytokines, together with Dan and DRM/Gremlin, represent a group of bone morphogenetic protein (BMP) antagonists that can bind directly to BMPs and inhibit their activity. (provided by RefSeq)

Immunogen

CER1 (AAH69491.1, 1 a.a. ~ 267 a.a) full-length human protein.

Sequence
MHLLLFQLLVLLPLGKTTRHQDGRQNQSSLSPVLLPRNQRELPTGNHEEAEEKPDLFVAVPHLVGTSPAGEGQRQREKMLSRFGRFWKKPEREMHPSRDSDSEPFPPGTQSLIQPIDGMKMEKSPLREEAKKFWHHFMFRKTPASQGVILPIKSHEVHWETCRTVPFSQTITHEGCEKVVVQNNLCFGKCGSVHFPGAAQHSHTSCSHCLPAKFTTMHLPLNCTELSSVIKVVMLVEECQCKVKTEHEDGHILHAGSQDSFIPGVSA

Biochem/physiol Actions

Cerberus 1 (CER1) is associated with the regulation of various pathways including nodal, Wnt, or BMP signaling pathways. During bone development, it acts as an inhibitor of BMPs (bone morphogenetic proteins). Polymorphisms in CER1 has link with osteoporosis, on bone mineral density (BMD) and bone markers in osteoporotic women. It has been reported that CER1 has been identified as a potential marker for bone mineral density and bone markers in osteoporotic women.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theodora Koromila et al.
Human genomics, 7, 21-21 (2013-10-22)
Osteoporosis has a multifactorial pathogenesis characterized by a combination of low bone mass and increased fragility. In our study, we focused on the effects of polymorphisms in CER1 and DKK1 genes, recently reported as important susceptibility genes for osteoporosis, on
Hidefumi Iwashita et al.
PloS one, 8(5), e64291-e64291 (2013-05-30)
To date, CXCR4 and E-cadherin double-positive cells detected by flow cytometry have been used to identify the differentiation of embryonic stem (ES) cells or induced pluripotent stem (iPS) cells into definitive endoderm (DE) lineages. Quantification of DE differentiation from ES/iPS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service