Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1402456

Sigma-Aldrich

Monoclonal Anti-OSMR antibody produced in mouse

clone 3E12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC150626, MGC150627, MGC75127, OSMRB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E12, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OSMR(9180)

General description

Oncostatin M receptor (OSMR) is a cell surface cytokine receptor. Oncostatin M and interleukin-31 are its ligands. The gene encoding this protein is localized on human chromosome 5p.

Immunogen

OSMR (NP_003990.1, 141 a.a. ~ 240 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QDSTGQDILFVFPKDKLVEEGTNVTICYVSRNIQNNVSCYLEGKQIHGEQLDPHVTAFNLNSVPFIRNKGTNIYCEASQGNVSEGMKGIVLFVSKVLEEP

Biochem/physiol Actions

Oncostatin M receptor (OSMR) acts as an activator of janus kinase/signal transducers and activators of transcription (JAK/STAT) and mitogen-activated protein kinase pathway. Thus, it has a role in stimulating cytokines and inflammatory substances. Mutation in the OSMR gene has been linked to primary localized cutaneous amyloidosis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of OSMR gene polymorphisms with rheumatoid arthritis and systemic lupus erythematosus patients.
Lin YZ
Autoimmunity (2014)
Overexpression of the oncostatin-M receptor in cervical squamous cell carcinoma is associated with epithelial-mesenchymal transition and poor overall survival.
Kucia-Tran JA
British Journal of Cancer (2016)
A novel missense mutation in oncostatin M receptor beta causing primary localized cutaneous amyloidosis.
Saeedi M
BioMed Research International (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service