Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1301739

Sigma-Aldrich

ANTI-HIST1H2BM (N-TERM) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

H2B/e, H2BFE, HIST1H2BM, Histone H2B type 1-M, Histone H2B.e

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
$683.00

$683.00


Please contact Customer Service for Availability


Select a Size

Change View
400 μL
$683.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$683.00


Please contact Customer Service for Availability

antibody form

IgG fraction of antiserum

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

13989 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
western blot: 1:250-1:500

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Shahid et al.
Proteomics, 18(20), e1800190-e1800190 (2018-09-21)
Lower urinary tract symptoms (LUTSs) are highly prevalent among the elderly and negatively impact quality of life. Since caffeinated beverages are enjoyed worldwide and the relationship between LUTS and caffeine is still not fully understood, it would be of particular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service