Skip to Content
MilliporeSigma
All Photos(1)

Documents

S7271

Supelco

Sample diffusion discs for LC columns

column I.D. 2.5 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.52

technique(s)

LPLC: suitable

column I.D.

2.5 cm

Looking for similar products? Visit Product Comparison Guide

General description

A 20 μm polyethylene disc sealed to an acrylic ring. Installed on top of column packing in an open-ended column, it protects the packing from disruption.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service